Anti-HNRNPAB Rabbit Recombinant Polyclonal Antibody (HRP (Horseradish Peroxidase))
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: HNRNPAB
Clonality: Recombinant Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77857-523
- 77740-230
- Antigen Symbol:
- HNRNPAB
- HNRNPAB
- Antigen Synonyms:
- ABBP 1|ABBP-1|ABBP1|Apobec 1 binding protein 1|APOBEC1 binding protein 1|APOBEC1-binding protein 1|Apolipoprotein B mRNA editing enzyme catalytic polypeptide 1- protein 1|FLJ40338|Heterogeneous nuclear ribonucleoprotein A/B|hnRNP A/B|hnRNP type A/B protein|HNRPAB|ROAA_HUMAN
- ABBP 1|ABBP-1|ABBP1|Apobec 1 binding protein 1|APOBEC1 binding protein 1|APOBEC1-binding protein 1|Apolipoprotein B mRNA editing enzyme catalytic polypeptide 1- protein 1|FLJ40338|Heterogeneous nuclear ribonucleoprotein A/B|hnRNP A/B|hnRNP type A/B protein|HNRPAB|ROAA_HUMAN
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- HRP (Horseradish Peroxidase)
- HRP (Horseradish Peroxidase)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q99729
- Q99729
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Immunogen:
- Recombinant Human Heterogeneous nuclear ribonucleoprotein A/B protein (1-68AA)
- Recombinant Human Heterogeneous nuclear ribonucleoprotein A/B protein (1-68AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice