Anti-HEPACAM Rabbit Recombinant Polyclonal Antibody (Biotinylated)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: HEPACAM
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77856-823
- 77739-631
- Antigen Symbol:
- HEPACAM
- HEPACAM
- Antigen Synonyms:
- FLJ25530|GlialCAM|HECAM_HUMAN|Hepatocyte cell adhesion molecule|Protein hepaCAM
- FLJ25530|GlialCAM|HECAM_HUMAN|Hepatocyte cell adhesion molecule|Protein hepaCAM
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Biotin
- Biotin
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q14CZ8
- Q14CZ8
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Immunogen:
- Recombinant Human Hepatocyte cell adhesion molecule protein (265-415AA)
- Recombinant Human Hepatocyte cell adhesion molecule protein (265-415AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice