Anti-LACTB Rabbit Polyclonal Antibody
77741-939
New Product
- Antigen Symbol:LACTB
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Mouse,Rat
- Western Blot:Yes
- Size:100 µl
- Form:Liquid
- Gene ID:P83111
- Antigen Synonyms:MRPL56| UNQ843/PRO1781Serine beta-lactamase-like protein mitochondrial| EC 3.4.-.-
- Storage Buffer:PBS, 0.1% sodium azide, 50% glycerol, pH 7.3
- Storage Temperature:Upon receipt, store at −20 or −80 °C, avoid repeated freeze
- Shipping Temperature:Shipped on Blue ice
- Immunogen:EC 3.4.-.-
- Purification:Antigen affinity purified
- Packaging:Vial
Type:
Antigen: LACTB
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse;Rat