Anti-ENTPD7 Rabbit Recombinant Polyclonal Antibody (HRP (Horseradish Peroxidase))
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: ENTPD7
Clonality: Recombinant Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77854-766
- 77735-950
- Antigen Symbol:
- ENTPD7
- ENTPD7
- Antigen Synonyms:
- DKFZp667O124|Ectonucleoside triphosphate diphosphohydrolase 7|ENTP7_HUMAN|ENTPD 7|LALP 1|LALP1|Lysosomal apyrase like protein 1|Lysosomal apyrase-like protein 1|NTPDase 7|NTPDase7
- DKFZp667O124|Ectonucleoside triphosphate diphosphohydrolase 7|ENTP7_HUMAN|ENTPD 7|LALP 1|LALP1|Lysosomal apyrase like protein 1|Lysosomal apyrase-like protein 1|NTPDase 7|NTPDase7
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- HRP (Horseradish Peroxidase)
- HRP (Horseradish Peroxidase)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q9NQZ7
- Q9NQZ7
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Immunogen:
- Recombinant human Ectonucleoside triphosphate diphosphohydrolase 7 protein (51-350AA)
- Recombinant human Ectonucleoside triphosphate diphosphohydrolase 7 protein (51-350AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice