Anti-AFF1 Rabbit Polyclonal Antibody
77739-528
New Product
- Antigen Symbol:AFF1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Mouse
- Form:Liquid
- Gene ID:P51825
- Antigen Synonyms:AF 4|AF 4 protein|AF4|AF4-MLL|AF4/FMR2 family member 1|AFF 1|AFF1|AFF1_HUMAN|ALL1 fused gene from chromosome 4|ALL1-fused gene from chromosome 4 protein|FEL|MGC134969|MLL/AF4|MLLT2|myeloid/lymphoid or mixed lineage leukemia (trithorax homolog|Drosophila)|translocated to|2|MYELOID/LYMPHOID OR MIXED LINEAGE LEUKEMIA TRANSLOCATED TO 2|PBM1|Pre B cell monocytic leukemia partner 1|Protein AF-4|Protein FEL|Proto-oncogene AF4
- Storage Buffer:PBS, 50% glycerol, 0.5% BSA, 0.02% sodium azide, pH 7.4
- Storage Temperature:Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:AF 4
- Purification:Antigen affinity purified
- Packaging:Vial
Recommended dilution: IHC:1:100 to 1:300, ELISA:1:20000
Type:
Antigen: AFF1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse