Anti-FABP5 Rabbit Polyclonal Antibody
77736-404
New Product
- Antigen Symbol:FABP5
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Mouse,Rat
- Western Blot:Yes
- Form:Liquid
- Gene ID:Q01469
- Antigen Synonyms:CFABP|Cutaneous fatty acid binding protein|DA11|Differentiation associated lipid binding protein LP2|E-FABP|EFABP|Epidermal-type fatty acid-binding protein|FABP5|FABP5_HUMAN|fatty acid binding protein 5 (psoriasis-associated)|Fatty acid binding protein epidermal|Fatty acid-binding protein 5|Fatty acid-binding protein|epidermal|Keratinocyte lipid binding protein|KFABP|Klbp|mal1|PA-FABP|PAFABP|Psoriasis associated fatty acid binding protein homolog|Psoriasis-associated fatty acid-binding protein homolog
- Storage Buffer:PBS, 0.1% sodium azide, 50% glycerol, pH 7.3
- Storage Temperature:Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:Psoriasis-associated fatty acid-binding protein homolog
- Purification:Antigen affinity purified
- Packaging:Vial
Type:
Antigen: FABP5
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse;Rat