Anti-OMA1 Rabbit Polyclonal Antibody
77745-465
New Product
- Antigen Symbol:OMA1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Mouse,Rat
- Western Blot:Yes
- Size:100 µl
- Form:Liquid
- Gene ID:Q96E52
- Antigen Synonyms:2010001O09Rik|DAB1|FLJ33782|Metalloendopeptidase Metalloendopeptidase mitochondrial|Metalloprotease-related protein 1|mitochondrial|MPRP 1|MPRP-1|MPRP1|OMA1 homolog zinc metallopeptidase|OMA1 homolog|zinc metallopeptidase (S. cerevisiae)|OMA1 zinc metallopeptidase|OMA1 zinc metallopeptidase homolog|OMA1_HUMAN|Overlapping activity with M-AAA protease|Overlapping with the m-AAA protease 1 homolog|peptidase|YKR087C|Zinc metallopeptidase ZMPOMA1
- Storage Buffer:PBS, 0.1% sodium azide, 50% glycerol, pH 7.3.
- Storage Temperature:Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:ZMPOMA1
- Purification:Antigen affinity purified
- Packaging:Vial
Type:
Antigen: OMA1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat