Anti-FOXL2 Rabbit Recombinant Polyclonal Antibody (Biotin)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: FOXL2
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77737-441
- 77855-561
- Antigen Symbol:
- FOXL2
- FOXL2
- Antigen Synonyms:
- Blepharophimosis|Blepharophimosis epicanthus inversus and ptosis 1|Blepharophimosis epicanthus inversus and ptosis|BPES 1|BPES|BPES1|Epicanthus inversus and ptosis 1|Forkhead box L2|Forkhead box protein L2|Forkhead transcription factor FOX L2|FOXL 2|FOXL2_HUMAN|PFRK|PINTO|PITUITARY FORKHEAD FACTOR|POF 3|POF3
- Blepharophimosis|Blepharophimosis epicanthus inversus and ptosis 1|Blepharophimosis epicanthus inversus and ptosis|BPES 1|BPES|BPES1|Epicanthus inversus and ptosis 1|Forkhead box L2|Forkhead box protein L2|Forkhead transcription factor FOX L2|FOXL 2|FOXL2_HUMAN|PFRK|PINTO|PITUITARY FORKHEAD FACTOR|POF 3|POF3
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Biotin
- Biotin
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- P58012
- P58012
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Immunogen:
- Recombinant human forkhead box protein L2 protein (116-216AA)
- Recombinant human Forkhead box protein L2 protein (116-216AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice