Anti-GNPAT Rabbit Recombinant Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: GNPAT
Clonality: Recombinant Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77738-632
- 77856-250
- Antigen Symbol:
- GNPAT
- Antigen Synonyms:
- Acyl CoA dihydroxyacetonephosphateacyltransferase|Acyl-CoA:dihydroxyacetonephosphateacyltransferase|DAP AT|DAP-AT|DAPAT|DHAP AT|DHAP-AT|DHAPAT|Dihydroxyacetone phosphate acyltransferase|EC 2.3.1.42|Glycerone-phosphate O-acyltransferase|Glyceronephosphate O acyltransferase|GNPAT|GNPAT_HUMAN|OTTHUMP00000036147
- Acyl CoA dihydroxyacetonephosphateacyltransferase|Acyl-CoA:dihydroxyacetonephosphateacyltransferase|DAP AT|DAP-AT|DAPAT|DHAP AT|DHAP-AT|DHAPAT|Dihydroxyacetone phosphate acyltransferase|EC 2.3.1.42|Glycerone-phosphate O-acyltransferase|Glyceronephosphate O acyltransferase|GNPAT|GNPAT_HUMAN|OTTHUMP00000036147
- Clonality:
- Recombinant Polyclonal
- Conjugation:
- FITC (Fluorescein Isothiocyanate)
- Reactivity:
- Human
- Host:
- Rabbit
- Gene ID:
- O15228
- O15228
- Isotype:
- IgG
- ELISA:
- Yes
- Immunogen:
- Recombinant Human Dihydroxyacetone phosphate acyltransferase protein (589-680AA)
- Recombinant Human Dihydroxyacetone phosphate acyltransferase protein (589-680AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at –20 °C or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 °C or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice