Anti-HIST1H2BD Rabbit Recombinant polyclonal Antibody (Biotinylated)
Supplier: AFG Bioscience
New Product
Certificates
About this item
0
Type:
Antigen: HIST1H2BD
Clonality: Recombinant polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77857-142
- 77739-873
- Antigen Symbol:
- HIST1H2BD
- HIST1H2BD
- Antigen Synonyms:
- H2BFB|HIRIP2|Histone H2B type 1-D|HIRA-interacting protein 2|Histone H2B.1 B|Histone H2B.b|H2B/b
- Histone H2B type 1-D|H2B/b|HIRIP2|H2BFB|HIRA-interacting protein 2|Histone H2B.1 B|Histone H2B.b
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Biotin
- Biotin
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- P58876
- P58876
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Immunogen:
- Recombinant Human Histone H2B type 1-D protein (2-57AA)
- Recombinant Human Histone H2B type 1-D protein (2-57AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at −20 or −80°C, avoid repeated freeze
- Upon receipt, store at −20 or −80 °C, avoid repeated freeze
- Shipping Temperature:
- Shipped on Blue ice
- Shipped on Blue ice