Anti-HIST1H4A Rabbit Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
Recommended dilution: IHC:1:10 to 1:100
Type:
Antigen: HIST1H4A
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77740-066
- 77857-425
- Antigen Symbol:
- HIST1H4A
- HIST1H4A
- Antigen Synonyms:
- dJ160A22.1|dJ160A22.2|dJ221C16.1|dJ221C16.9|FO108|H4|H4.k|H4/a|H4/b|H4/c|H4/d|H4/e|H4/g|H4/h|H4/I|H4/j|H4/k|H4/m|H4/n|H4/p|H4_HUMAN|H4F2|H4F2iii|H4F2iv|H4FA|H4FB|H4FC|H4FD|H4FE|H4FG|H4FH|H4FI|H4FJ|H4FK|H4FM|H4FN|H4M|HIST1H4A|HIST1H4B|HIST1H4C|HIST1H4D|HIST1H4E|HIST1H4F|HIST1H4H|HIST1H4I|HIST1H4J|HIST1H4K|HIST1H4L|HIST2H4|HIST2H4A|Hist4h4|Histone 1 H4a|Histone 1 H4b|Histone 1 H4c|Histone 1 H4d|Histone 1 H4e|Histone 1 H4f|Histone 1 H4h|Histone 1 H4i|Histone 1 H4j|Histone 1 H4k|Histone 1 H4l|Histone 2 H4a|histone 4 H4|Histone H4|MGC24116
- dJ160A22.1| dJ160A22.2| dJ221C16.1| dJ221C16.9| FO108| H4| H4.k| H4/a| H4/b| H4/c| H4/d| H4/e| H4/g| H4/h| H4/I| H4/j| H4/k| H4/m| H4/n| H4/p| H4_HUMAN| H4F2| H4F2iii| H4F2iv| H4FA| H4FB| H4FC| H4FD| H4FE| H4FG| H4FH| H4FI| H4FJ| H4FK| H4FM| H4FN| H4M| HIST1H4A| HIST1H4B| HIST1H4C| HIST1H4D| HIST1H4E| HIST1H4F| HIST1H4H| HIST1H4I| HIST1H4J| HIST1H4K| HIST1H4L| HIST2H4| HIST2H4A| Hist4h4| Histone 1 H4a| Histone 1 H4b| Histone 1 H4c| Histone 1 H4d| Histone 1 H4e| Histone 1 H4f| Histone 1 H4h| Histone 1 H4i| Histone 1 H4j| Histone 1 H4k| Histone 1 H4l| Histone 2 H4a| histone 4 H4| Histone H4| MGC24116
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- P62805
- P62805
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Immunogen:
- Peptide sequence around site of Lys (91) derived from Human Histone H4
- Peptide sequence around site of Lys (91) derived from Human Histone H4
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice