Anti-GJB6 Rabbit Recombinant Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
Recommended dilution: IHC: 1:20 to 1:200, IF: 1:50 to 1:200
Type:
Antigen: GJB6
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77738-320
- 77856-040
- Antigen Symbol:
- GJB6
- Antigen Synonyms:
- Connexin 30|Connexin-30|Cx30|CXB6_HUMAN|DFNA3|DFNA3B|DFNB1B|ECTD2|ectodermal dysplasia 2|hidrotic (Clouston syndrome)|ED2|EDH|Gap junction beta 6 protein|Gap junction beta-6 protein|gap junction protein|beta 6 (connexin 30)|beta 6|Gap junction protein|30kDa|GJB6|HED|HED2|NSRD1
- Connexin 30|Connexin-30|Cx30|CXB6_HUMAN|DFNA3|DFNA3B|DFNB1B|ECTD2|ectodermal dysplasia 2|hidrotic (Clouston syndrome)|ED2|EDH|Gap junction beta 6 protein|Gap junction beta-6 protein|gap junction protein|beta 6 (connexin 30)|beta 6|Gap junction protein|30kDa|GJB6|HED|HED2|NSRD1
- Clonality:
- Recombinant Polyclonal
- Conjugation:
- Unconjugated
- Reactivity:
- Human
- Host:
- Rabbit
- Gene ID:
- O95452
- O95452
- Isotype:
- IgG
- ELISA:
- Yes
- ImmunoChemistry:
- Yes
- ImmunoFluorescence:
- Yes
- Immunogen:
- Recombinant Human Gap junction beta-6 protein (216-261AA)
- Recombinant Human Gap junction beta-6 protein (216-261AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at –20 °C or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 °C or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice