Anti-HCRTR1 Rabbit Recombinant Polyclonal Antibody (HRP (Horseradish Peroxidase))
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: HCRTR1
Clonality: Recombinant Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77739-561
- 77856-788
- Antigen Symbol:
- HCRTR1
- HCRTR1
- Antigen Synonyms:
- HCRTR 1|Hypocretin receptor type 1|Orexin Receptor 1|Orexin receptor type 1|Ox-1-R|Ox1-R|Ox1r|OX1R_HUMAN
- HCRTR 1|Hypocretin receptor type 1|Orexin Receptor 1|Orexin receptor type 1|Ox-1-R|Ox1-R|Ox1r|OX1R_HUMAN
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- HRP (Horseradish Peroxidase)
- HRP (Horseradish Peroxidase)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- O43613
- O43613
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Immunogen:
- Recombinant Human Orexin receptor type 1 protein (1-46AA)
- Recombinant Human Orexin receptor type 1 protein (1-46AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice