Anti-GAB1 Rabbit Polyclonal Antibody
Catalog # 77737-006
Supplier: AFG Bioscience
New Product
Specifications
- Antigen Symbol:GAB1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Mouse
- Size:50 µg
- Form:Liquid
- Gene ID:Q13480
- Antigen Synonyms:Gab 1|GAB1_HUMAN|GRB 2 associated binder 1|GRB 2 associated binding protein 1|GRB2 associated binding protein 1 isoform a|GRB2 associated binding protein 1 isoform b|GRB2-associated binder 1|GRB2-associated-binding protein 1|Growth factor receptor bound protein 2-associated protein 1
- Storage Buffer:PBS, 50% glycerol, 0.5% BSA, 0.02% sodium azide, pH 7.4.
- Storage Temperature:Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:Gab 1
- Purification:Antigen affinity purified
- Packaging:Vial
Specifications
About this item
Recommended dilution: IHC: 1:100 to 1:300, ELISA: 1:5000
Type:
Antigen: GAB1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse