Anti-HEXA Rabbit Polyclonal Antibody
Catalog # 77739-650
Supplier: AFG Bioscience
New Product
Specifications
- Antigen Symbol:HEXA
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Mouse,Rat
- Western Blot:Yes
- Size:100 µl
- Form:Liquid
- Gene ID:P06865
- Antigen Synonyms:Beta hexosaminidase alpha chain precursor| Beta hexosaminidase subunit alpha| Beta N acetylhexosaminidase| Beta N acetylhexosaminidase subunit alpha| Beta-hexosaminidase A| Beta-hexosaminidase subunit alpha| Beta-N-acetylhexosaminidase subunit alpha| HEXA_HUMAN| Hexosaminidase A (alpha polypeptide)| Hexosaminidase A alpha polypeptide| Hexosaminidase A| Hexosaminidase subunit A| MGC99608| N acetyl beta glucosaminidase| N acetyl beta glucosaminidase subunit alpha| N-acetyl-beta-glucosaminidase subunit alpha| TSD
- Storage Buffer:PBS, 0.1% sodium azide, 50% glycerol, pH 7.3
- Storage Temperature:Upon receipt, store at −20 or −80 °C, avoid repeated freeze
- Shipping Temperature:Shipped on Blue ice
- Immunogen:TSD
- Purification:Antigen affinity purified
- Packaging:Vial
Specifications
About this item
Type:
Antigen: HEXA
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse;Rat