Anti-ELF3 Rabbit Recombinant Polyclonal Antibody (HRP (Horseradish Peroxidase))
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: ELF3
Clonality: Recombinant Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77854-673
- 77735-801
- Antigen Symbol:
- ELF3
- ELF3
- Antigen Synonyms:
- E74 like factor 3|E74 like factor 3 ets domain transcription factor|E74 like factor 3 ets domain transcription factor epithelial specific|E74 like factor 3 ETS domain transcription factor serine box epithelial specific|E74-like factor 3|ELF3_HUMAN|Epithelial restricted with serine box|Epithelial-restricted with serine box|Epithelium restricted Ets protein ESX|Epithelium specific Ets factor 1|Epithelium specific Ets transcription factor 1|Epithelium-restricted Ets protein ESX|Epithelium-specific Ets transcription factor 1|EPR 1|EPR1|ERT|ESE-1|ESX|Ets domain transcription factor serine box|Ets domain transcription factor serine box epithelial specific|Ets transcription factor|ETS-related transcription factor Elf-3|jen|MGC139318
- E74 like factor 3|E74 like factor 3 ets domain transcription factor|E74 like factor 3 ets domain transcription factor epithelial specific|E74 like factor 3 ETS domain transcription factor serine box epithelial specific|E74-like factor 3|ELF3_HUMAN|Epithelial restricted with serine box|Epithelial-restricted with serine box|Epithelium restricted Ets protein ESX|Epithelium specific Ets factor 1|Epithelium specific Ets transcription factor 1|Epithelium-restricted Ets protein ESX|Epithelium-specific Ets transcription factor 1|EPR 1|EPR1|ERT|ESE-1|ESX|Ets domain transcription factor serine box|Ets domain transcription factor serine box epithelial specific|Ets transcription factor|ETS-related transcription factor Elf-3|jen|MGC139318
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- HRP (Horseradish Peroxidase)
- HRP (Horseradish Peroxidase)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- P78545
- P78545
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Immunogen:
- Recombinant Human ETS-related transcription factor Elf-3 protein (147-371AA)
- Recombinant Human ETS-related transcription factor Elf-3 protein (147-371AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice