Anti-OCIAD1 Rabbit Polyclonal Antibody
Catalog # 77745-336
Supplier: AFG Bioscience
New Product
Specifications
- Antigen Symbol:OCIAD1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human,Mouse,Rat
- Western Blot:Yes
- Size:100 µl
- Form:Liquid
- Gene ID:Q9NX40
- Antigen Synonyms:OCAD1_HUMAN|Ovarian carcinoma immunoreactive antigen|MGC111072|OCIA domain-containing protein 1|Asrij|OCIA|FLJ20455|OCIA domain containing 1|TPA018
- Storage Buffer:PBS, 0.1% sodium azide, 50% glycerol, pH 7.3
- Storage Temperature:Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:TPA018
- Purification:Antigen affinity purified
- Packaging:Vial
Specifications
About this item
Type:
Antigen: OCIAD1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse;Rat