Anti-MAPKAP1 Rabbit Polyclonal Antibody
77742-767
New Product
- Antigen Symbol:MAPKAP1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human,Mouse,Rat
- Western Blot:Yes
- Size:100 µl
- Form:Liquid
- Gene ID:Q9BPZ7
- Antigen Synonyms:MAPKAP 1|MAPKAP1|MEKK2 interacting protein 1|MGC2745|MIP 1|MIP1|Mitogen activated protein kinase associated protein 1|Mitogen-activated protein kinase 2-associated protein 1|mSIN1|OTTHUMP00000064207|Ras inhibitor MGC2745|SAPK interacting protein 1|SAPK-interacting protein 1|SIN 1|SIN1_HUMAN|SIN1b|SIN1g|Stress activated map kinase interacting protein 1|Stress activated protein kinase interacting 1|Stress-activated map kinase-interacting protein 1|Target of rapamycin complex 2 subunit MAPKAP1|TORC2 subunit MAPKAP1
- Storage Buffer:PBS, 0.1% sodium azide, 50% glycerol, pH 7.3
- Storage Temperature:Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:TORC2 subunit MAPKAP1
- Purification:Antigen affinity purified
- Packaging:Vial
Type:
Antigen: MAPKAP1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse;Rat