Anti-GUCY2D Rabbit Recombinant Polyclonal Antibody (Biotinylated)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: GUCY2D
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77739-289
- 77856-629
- Antigen Symbol:
- GUCY2D
- GUCY2D
- Antigen Synonyms:
- CORD6|GUC1A4|GUC2D|RETGC|RETGC1|Retinal guanylyl cyclase 1|RETGC-1|EC 4.6.1.2|CG-E|Guanylate cyclase 2D|retinal|Rod outer segment membrane guanylate cyclase|ROS-GC
- CORD6|GUC1A4|GUC2D|RETGC|RETGC1|Retinal guanylyl cyclase 1|RETGC-1|EC 4.6.1.2|CG-E|Guanylate cyclase 2D|retinal|Rod outer segment membrane guanylate cyclase|ROS-GC
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Biotin
- Biotin
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q02846
- Q02846
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Immunogen:
- Recombinant Human Retinal guanylyl cyclase 1 protein (362-462AA)
- Recombinant Human Retinal guanylyl cyclase 1 protein (362-462AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- 0.03% proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice