Anti-IL1RAPL1 Rabbit Recombinant Polyclonal Antibody (Biotinylated)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: IL1RAPL1
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77740-983
- 77858-039
- Antigen Symbol:
- IL1RAPL1
- IL1RAPL1
- Antigen Synonyms:
- IL 1 RAPL 1|IL 1RAPL 1|IL-1-RAPL-1|IL-1RAPL-1|IL1R8|IL1RAPL 1|IL1RAPL|IL1RAPL-1|Interleukin 1 receptor 8|Interleukin 1 receptor accessory protein like 1|Interleukin-1 receptor accessory protein-like 1|IRPL1_HUMAN|Mental retardation X linked 10|Mental retardation X linked 21|Mental retardation X linked 34|MRX 21|MRX 34|MRX10|MRX21|MRX34|Oligophrenin 4|Oligophrenin-4|Oligophrenin4|OPHN 4|OPHN4|Three immunoglobulin domain containing IL 1 receptor related 2|Three immunoglobulin domain-containing IL-1 receptor-related 2|TIGIRR 2|TIGIRR-2|TIGIRR2|X linked interleukin 1 receptor accessory protein like 1|X-linked interleukin-1 receptor accessory protein-like 1
- IL 1 RAPL 1|IL 1RAPL 1|IL-1-RAPL-1|IL-1RAPL-1|IL1R8|IL1RAPL 1|IL1RAPL|IL1RAPL-1|Interleukin 1 receptor 8|Interleukin 1 receptor accessory protein like 1|Interleukin-1 receptor accessory protein-like 1|IRPL1_HUMAN|Mental retardation X linked 10|Mental retardation X linked 21|Mental retardation X linked 34|MRX 21|MRX 34|MRX10|MRX21|MRX34|Oligophrenin 4|Oligophrenin-4|Oligophrenin4|OPHN 4|OPHN4|Three immunoglobulin domain containing IL 1 receptor related 2|Three immunoglobulin domain-containing IL-1 receptor-related 2|TIGIRR 2|TIGIRR-2|TIGIRR2|X linked interleukin 1 receptor accessory protein like 1|X-linked interleukin-1 receptor accessory protein-like 1
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Biotin
- Biotin
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q9NZN1
- Q9NZN1
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Immunogen:
- Recombinant Human Interleukin-1 receptor accessory protein-like 1 protein (564-679AA)
- Recombinant Human Interleukin-1 receptor accessory protein-like 1 protein (564-679AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- 0.03% proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together