Anti-EXOSC4 Rabbit Polyclonal Antibody
77736-300
New Product
- Antigen Symbol:EXOSC4
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human,Mouse,Rat
- Western Blot:Yes
- Size:100 µl
- Form:Liquid
- Gene ID:Q9NPD3
- Antigen Synonyms:EXOS4_HUMAN|EXOSC 4|EXOSC4|Exosome complex component RRP41|Exosome complex exonuclease RRP41|Exosome component 4|Exosome component Rrp41|FLJ20591|hRrp41p|p12A|Ribosomal RNA processing protein 41|Ribosomal RNA-processing protein 41|RRP 41|RRP41|RRP41A|Rrp41p|SKI 6|SKI6|Ski6p
- Storage Buffer:PBS, 0.1% sodium azide, 50% glycerol, pH 7.3.
- Storage Temperature:Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:Ribosomal RNA processing protein 41
- Purification:Antigen affinity purified
- Packaging:Vial
Type:
Antigen: EXOSC4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat