Anti-HSD3B1 Rabbit Recombinant Polyclonal Antibody (HRP (Horseradish Peroxidase))
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: HSD3B1
Clonality: Recombinant Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77857-691
- 77740-434
- Antigen Symbol:
- HSD3B1
- HSD3B1
- Antigen Synonyms:
- 3 beta and steroid delta isomerase 1|3 beta and steroid hydroxy delta 5 steroid dehydrogenase|3 beta HSD I|3 beta-hydroxysteroid dehydrogenase/Delta 5-->,4-isomerase type I|3-beta-HSD I|3-beta-hydroxy-5-ene steroid dehydrogenase|3BETAHSD|3BH|3BHS1_HUMAN|3BHSD|Delta-5-3-ketosteroid isomerase|HGNC:5217|HSD3B|HSDB3|HSDB3A|Hydroxy delta 5 steroid dehydrogenase 3 beta and steroid delta isomerase 1|Hydroxy delta 5 steroid dehydrogenase|Progesterone reductase|SDR11E1|Steroid Delta-isomerase|trophoblast antigen|Trophoblast antigen FDO161G
- 3 beta and steroid delta isomerase 1|3 beta and steroid hydroxy delta 5 steroid dehydrogenase|3 beta HSD I|3 beta-hydroxysteroid dehydrogenase/Delta 5-->,4-isomerase type I|3-beta-HSD I|3-beta-hydroxy-5-ene steroid dehydrogenase|3BETAHSD|3BH|3BHS1_HUMAN|3BHSD|Delta-5-3-ketosteroid isomerase|HGNC:5217|HSD3B|HSDB3|HSDB3A|Hydroxy delta 5 steroid dehydrogenase 3 beta and steroid delta isomerase 1|Hydroxy delta 5 steroid dehydrogenase|Progesterone reductase|SDR11E1|Steroid Delta-isomerase|trophoblast antigen|Trophoblast antigen FDO161G
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- HRP (Horseradish Peroxidase)
- HRP (Horseradish Peroxidase)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- P14060
- P14060
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Immunogen:
- Recombinant human 3 beta-hydroxysteroid dehydrogenase/Delta 5 - >4-isomerase type 1 protein (127-267AA)
- Recombinant human 3 beta-hydroxysteroid dehydrogenase/Delta 5-->4-isomerase type 1 protein (127-267AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice