Anti-GJA5 Rabbit Recombinant Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: GJA5
Clonality: Recombinant Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77738-298
- 77856-018
- Antigen Symbol:
- GJA5
- GJA5
- Antigen Synonyms:
- Connexin 40|Connexin 40 / Connexin-40|Cx40|CXA5_HUMAN|Gap junction alpha 5 protein|Gap junction alpha-5 protein|Gap junction protein alpha 5 40kD (connexin 40)|Gap junction protein alpha 5|MGC11185
- Connexin 40|Connexin 40 / Connexin-40|Cx40|CXA5_HUMAN|Gap junction alpha 5 protein|Gap junction alpha-5 protein|Gap junction protein alpha 5 40kD (connexin 40)|Gap junction protein alpha 5|MGC11185
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- FITC (Fluorescein Isothiocyanate)
- FITC (Fluorescein Isothiocyanate)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- P36382
- P36382
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Immunogen:
- Recombinant Human Gap junction alpha-5 protein (227-358AA)
- Recombinant Human Gap junction alpha-5 protein (227-358AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, protein G purified
- >95%, protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice