Anti-FGF12 Rabbit Recombinant Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
Recommended dilution: IHC: 1:20 to 1:200
Type:
Antigen: FGF12
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77855-331
- 77737-100
- Antigen Symbol:
- FGF12
- FGF12
- Antigen Synonyms:
- EIEE47|FGF-12|FGF12_HUMAN|FGF12B|FHF-1|FHF1|Fibroblast growth factor 12|Fibroblast growth factor 12B|Fibroblast growth factor FGF 12b|Fibroblast growth factor homologous factor 1|Myocyte activating factor|Myocyte-activating factor
- EIEE47|FGF-12|FGF12_HUMAN|FGF12B|FHF-1|FHF1|Fibroblast growth factor 12|Fibroblast growth factor 12B|Fibroblast growth factor FGF 12b|Fibroblast growth factor homologous factor 1|Myocyte activating factor|Myocyte-activating factor
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- P61328
- P61328
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Immunogen:
- Recombinant human Fibroblast growth factor 12 protein (1-181AA)
- Recombinant human Fibroblast growth factor 12 protein (1-181AA)
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage Buffer:
- PBS, 0.02% sodium azide, 50% glycerol, pH 7.3.
- PBS, 0.02% sodium azide, 50% glycerol, pH 7.3.
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice