Anti-GJA8 Rabbit Recombinant Polyclonal Antibody (Biotinylated)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: GJA8
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77856-023
- 77738-302
- Antigen Symbol:
- GJA8
- GJA8
- Antigen Synonyms:
- CAE|CAE1|cataract|zonular pulverulent 1 (FY-linked)|Connexin 50|Connexin-50|Cx50|CXA8_HUMAN|Gap junction alpha 8 protein|Gap junction alpha-8 protein|Gap junction membrane channel protein alpha 8 (connexin 50)|Gap junction protein alpha 8|gap junction protein|alpha 8|50kD|Lens fiber protein MP70
- CAE|CAE1|cataract|zonular pulverulent 1 (FY-linked)|Connexin 50|Connexin-50|Cx50|CXA8_HUMAN|Gap junction alpha 8 protein|Gap junction alpha-8 protein|Gap junction membrane channel protein alpha 8 (connexin 50)|Gap junction protein alpha 8|gap junction protein|alpha 8|50kD|Lens fiber protein MP70
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Biotin
- Biotin
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- P48165
- P48165
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Immunogen:
- Recombinant Human Gap junction alpha-8 protein (286-432AA)
- Recombinant Human Gap junction alpha-8 protein (286-432AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice