Anti-EPX Rabbit Recombinant Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
Recommended dilution: IHC:1:200 to 1:500
Type:
Antigen: EPX
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77736-062
- 77854-857
- Antigen Symbol:
- EPX
- Antigen Synonyms:
- Eosinophil peroxidase [Precursor]|Eosinophil peroxidase|Eosinophil peroxidase heavy chain|EPER|EPO|EPP|EPX|EPX PEN|PERE|PERE_HUMAN
- Eosinophil peroxidase [Precursor]|Eosinophil peroxidase|Eosinophil peroxidase heavy chain|EPER|EPO|EPP|EPX|EPX PEN|PERE|PERE_HUMAN
- Clonality:
- Recombinant Polyclonal
- Conjugation:
- Unconjugated
- Reactivity:
- Human
- Host:
- Rabbit
- Gene ID:
- P11678
- P11678
- Isotype:
- IgG
- ELISA:
- Yes
- ImmunoChemistry:
- Yes
- Immunogen:
- Recombinant Human Eosinophil peroxidase protein (140-300AA)
- Recombinant Human Eosinophil peroxidase protein (140-300AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95% Protein G purified
- >95% Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice