Anti-NCK1 Rabbit Recombinant Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
Recommended dilution: WB: 1:500 to 1:2000, IHC: 1:20 to 1:200
Type:
Antigen: NCK1
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Rat
Specifications
- Catalog No:
- 77744-012
- 77744-013
- Antigen Symbol:
- NCK1
- NCK1
- Antigen Synonyms:
- NCK tyrosine kinase|Nck-1|Cytoplasmic protein NCK1_HUMAN|NCK adaptor protein 1|Melanoma Nck protein|NCKalpha|NCK 1|SH2/SH3 adaptor protein NCK-alpha|SH2/SH3 adaptor protein NCK alpha|NCK alpha|NCK|Non catalytic region of tyrosine kinase|MGC12668|NCK1
- NCK tyrosine kinase|Nck-1|Cytoplasmic protein NCK1_HUMAN|NCK adaptor protein 1|Melanoma Nck protein|NCKalpha|NCK 1|SH2/SH3 adaptor protein NCK-alpha|SH2/SH3 adaptor protein NCK alpha|NCK alpha|NCK|Non catalytic region of tyrosine kinase|MGC12668|NCK1
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human, Rat
- Human, Rat
- Host:
- Rabbit
- Rabbit
- Gene ID:
- P16333
- P16333
- Isotype:
- IgG
- IgG
- Western Blot:
- Yes
- Yes
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Immunogen:
- Cytoplasmic protein NCK1 protein (58-377AA)
- Cytoplasmic protein NCK1 protein (58-377AA)
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage Buffer:
- PBS, 0.02% sodium azide, 50% glycerol, pH 7.3
- PBS, 0.02% sodium azide, 50% glycerol, pH 7.3
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice