Anti-NDUFA3 Rabbit Recombinant Polyclonal Antibody (Biotinylated)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: NDUFA3
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77744-139
- 77744-140
- Antigen Symbol:
- NDUFA3
- NDUFA3
- Antigen Synonyms:
- NADH dehydrogenase [ubiquinone] 1 alpha subcomplex subunit 3|Complex I-B9|CI-B9|NADH-ubiquinone oxidoreductase B9 subunit
- NADH dehydrogenase [ubiquinone] 1 alpha subcomplex subunit 3|Complex I-B9|CI-B9|NADH-ubiquinone oxidoreductase B9 subunit
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Biotin
- Biotin
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- O95167
- O95167
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Immunogen:
- NADH dehydrogenase [ubiquinone] 1 alpha subcomplex subunit 3 protein (2-84AA)
- NADH dehydrogenase [ubiquinone] 1 alpha subcomplex subunit 3 protein (2-84AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice