Order Entry
Puerto Rico
ContactUsLinkComponent
Anti-HAL Rabbit Recombinant polyclonal Antibody
Anti-HAL Rabbit Recombinant polyclonal Antibody
Supplier:  AFG Bioscience
New Product
Certificates
undefined
Anti-HAL Rabbit Recombinant polyclonal Antibody
Supplier:  AFG Bioscience
New Product

Specifications

  • Catalog No:
  • 77856-707
  • 77739-402
  • Antigen Symbol:
  • HAL
  • HAL
  • Antigen Synonyms:
  • dy| hutH 1|HAL protein|Histidase|Histidine ammonia lyase|Histidine ammonia-lyase|HSTD|HutH|HUTH_HUMAN
  • dy| hutH 1| HAL protein| Histidase| Histidine ammonia lyase| Histidine ammonia-lyase| HSTD| HutH| HUTH_HUMAN
  • Clonality:
  • Recombinant Polyclonal
  • Recombinant Polyclonal
  • Conjugation:
  • Unconjugated
  • Unconjugated
  • Reactivity:
  • Human
  • Human
  • Host:
  • Rabbit
  • Rabbit
  • Gene ID:
  • P42357
  • P42357
  • Isotype:
  • IgG
  • IgG
  • ELISA:
  • Yes
  • Yes
  • ImmunoChemistry:
  • Yes
  • Yes
  • ImmunoFluorescence:
  • Yes
  • Yes
  • Immunogen:
  • Recombinant Human Histidine ammonia-lyase protein (36-152AA)
  • Recombinant Human Histidine ammonia-lyase protein (36-152AA)
  • Form:
  • Liquid
  • Liquid
  • Purification:
  • >95%, Protein G purified
  • >95%, Protein G purified
  • Storage Buffer:
  • 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
  • 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
  • Storage Temperature:
  • Upon receipt, store at −20 or −80°C, avoid repeated freeze
  • Upon receipt, store at −20 or −80 °C, avoid repeated freeze
  • Shipping Temperature:
  • Shipped on Blue ice
  • Shipped on Blue ice

Specifications