Anti-NOXA1 Rabbit Recombinant Polyclonal Antibody (Biotinylated)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: NOXA1
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77744-742
- 77744-741
- Antigen Symbol:
- NOXA1
- NOXA1
- Antigen Synonyms:
- Antigen NY CO 31|Antigen NY-CO-31|FLJ25475|Inhibitory NADPH oxidase activator 1|MGC131800|NADPH oxidase activator 1|NCF2 like protein|NCF2-like protein|NOX activator 1|NOXA 1|Noxa1|NOXA1_HUMAN|NY CO 31|p51 nox|p51-nox|p51NOX|P67phox like factor|P67phox-like factor|SDCCAG31|Serologically defined colon cancer antigen 31
- Antigen NY CO 31|Antigen NY-CO-31|FLJ25475|Inhibitory NADPH oxidase activator 1|MGC131800|NADPH oxidase activator 1|NCF2 like protein|NCF2-like protein|NOX activator 1|NOXA 1|Noxa1|NOXA1_HUMAN|NY CO 31|p51 nox|p51-nox|p51NOX|P67phox like factor|P67phox-like factor|SDCCAG31|Serologically defined colon cancer antigen 31
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Biotin
- Biotin
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q86UR1
- Q86UR1
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Immunogen:
- NADPH oxidase activator 1 protein (150-313AA)
- NADPH oxidase activator 1 protein (150-313AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at –20 °C or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 °C or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice