Anti-FUCA1 Rabbit Recombinant Polyclonal Antibody (HRP (Horseradish Peroxidase))
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: FUCA1
Clonality: Recombinant Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77737-710
- 77855-646
- Antigen Symbol:
- FUCA1
- FUCA1
- Antigen Synonyms:
- Alpha L fucosidase I|Alpha L fucoside fucohydrolase|Alpha-L-fucosidase 1|Alpha-L-fucosidase I|Alpha-L-fucoside fucohydrolase 1|FUCA|FUCO_HUMAN|fucosidase|alpha-L|tissue|alpha-L- 1|Tissue alpha L fucosidase|Tissue alpha-L-fucosidase
- Alpha L fucosidase I|Alpha L fucoside fucohydrolase|Alpha-L-fucosidase 1|Alpha-L-fucosidase I|Alpha-L-fucoside fucohydrolase 1|FUCA|FUCO_HUMAN|fucosidase|alpha-L|tissue|alpha-L- 1|Tissue alpha L fucosidase|Tissue alpha-L-fucosidase
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- HRP (Horseradish Peroxidase)
- HRP (Horseradish Peroxidase)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- P04066
- P04066
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Immunogen:
- Recombinant human Tissue alpha-L-fucosidase protein (32-158AA)
- Recombinant human Tissue alpha-L-fucosidase protein (32-158AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice