Anti-PRLHR Rabbit Polyclonal Antibody
Catalog # 77737-542
Supplier: AFG Bioscience
New Product
Specifications
- Antigen Symbol:PRLHR
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human,Mouse,Rat
- Western Blot:Yes
- Size:50 µg
- Form:Liquid
- Gene ID:P49683
- Antigen Synonyms:G protein coupled receptor 10|G-protein coupled receptor 10|GPR 10|GPR10|GR 3|GR3|hGR 3|hGR3|MGC126539|MGC126541|OTTHUMP00000020584|PRLH RECEPTOR|PRLHR_HUMAN|Prolactin releasing hormone receptor|Prolactin releasing peptide receptor|Prolactin-releasing peptide receptor|PrRP receptor|PrRPR|uhr-1
- Storage Buffer:PBS, 50% glycerol, 0.5% BSA, 0.02% sodium azide, pH 7.4
- Storage Temperature:Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:G protein coupled receptor 10
- Purification:Antigen affinity purified
- Packaging:Vial
Specifications
About this item
Recommended dilution: WB: 1:500 to 1:2000, IF: 1:200 to 1:1000, ELISA: 1:20000
Type:
Antigen: PRLHR
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse;Rat