Anti-GIMAP5 Rabbit Polyclonal Antibody
Catalog # 77738-259
Supplier: AFG Bioscience
New Product
Specifications
- Antigen Symbol:GIMAP5
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human
- Western Blot:Yes
- Size:100 µl
- Form:Liquid
- Gene ID:Q96F15
- Antigen Synonyms:FLJ11296|GIMA5_HUMAN|GIMAP 5|GTPase IMAP family member 5|hIAN5|HIMAP3|IAN 5|IAN4|IAN4L1|IAN5|IMAP3|Immune associated nucleotide 4 like 1|Immunity associated nucleotide 4 like 1 protein|Immunity associated protein 3
- Storage Buffer:PBS, 0.1% sodium azide, 50% glycerol, pH 7.3
- Storage Temperature:Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:Immunity associated protein 3
- Purification:Antigen affinity purified
- Packaging:Vial
Specifications
About this item
Type:
Antigen: GIMAP5
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human