Anti-HSD11B1 Rabbit Recombinant Polyclonal Antibody
New Product
Recommended dilution: IHC: 1:20 to 1:200
Type:
Antigen: HSD11B1
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
- Catalog No:
- 77740-405
- 77857-669
- Antigen Symbol:
- HSD11B1
- HSD11B1
- Antigen Synonyms:
- 11 beta HSD 1|11 beta HSD1|11 beta hydroxysteroid dehydrogenase 1|11 DH|11-beta hydroxysteroid dehydrogenase|type 1|11-beta-HSD1|11-beta-hydroxysteroid dehydrogenase 1|11-DH|11DH|Corticosteroid 11 beta dehydrogenase isozyme 1|Corticosteroid 11-beta-dehydrogenase isozyme 1|CORTRD2|DHI1_HUMAN|HDL|HSD 11|HSD11|HSD11B|HSD11L|Hydroxysteroid (11 beta) dehydrogenase|Hydroxysteroid (11 beta) dehydrogenase 1|MGC13539|SDR26C1|short chain dehydrogenase/reductase family 26C|member 1
- 11 beta HSD 1|11 beta HSD1|11 beta hydroxysteroid dehydrogenase 1|11 DH|11-beta hydroxysteroid dehydrogenase|type 1|11-beta-HSD1|11-beta-hydroxysteroid dehydrogenase 1|11-DH|11DH|Corticosteroid 11 beta dehydrogenase isozyme 1|Corticosteroid 11-beta-dehydrogenase isozyme 1|CORTRD2|DHI1_HUMAN|HDL|HSD 11|HSD11|HSD11B|HSD11L|Hydroxysteroid (11 beta) dehydrogenase|Hydroxysteroid (11 beta) dehydrogenase 1|MGC13539|SDR26C1|short chain dehydrogenase/reductase family 26C|member 1
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- P28845
- P28845
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Immunogen:
- Recombinant Human Corticosteroid 11-beta-dehydrogenase isozyme 1 protein (25-292AA)
- Recombinant Human Corticosteroid 11-beta-dehydrogenase isozyme 1 protein (25-292AA)
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage Buffer:
- PBS, 0.02% sodium azide, 50% glycerol, pH 7.3.
- PBS, 0.02% sodium azide, 50% glycerol, pH 7.3.
- Storage Temperature:
- Upon receipt, store at –20 °C or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 °C or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice