Anti-KCTD5 Rabbit Polyclonal Antibody
77741-537
New Product
- Antigen Symbol:KCTD5
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human,Mouse,Rat
- Western Blot:Yes
- Size:100 µl
- Form:Liquid
- Gene ID:Q9NXV2
- Antigen Synonyms:FLJ20040|BTB/POZ domain-containing protein KCTD5_HUMAN|Potassium channel tetramerisation domain containing 5|KCTD5
- Storage Buffer:PBS, 0.1% sodium azide, 50% glycerol, pH 7.3
- Storage Temperature:Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:Potassium channel tetramerisation domain containing 5
- Purification:Antigen affinity purified
- Packaging:Vial
Type:
Antigen: KCTD5
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse;Rat