Anti-MGRN1 Rabbit Polyclonal Antibody
77743-104
New Product
- Antigen Symbol:MGRN1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Mouse,Rat
- Western Blot:Yes
- Form:Liquid
- Gene ID:O60291
- Antigen Synonyms:E3 ubiquitin-protein ligase MGRN1|KIAA0544|Mahogunin ring finger 1|Mahogunin RING finger protein 1|MGRN 1|Mgrn1|MGRN1_HUMAN|Probable E3 ubiquitin protein ligase MGRN1|RING finger protein 156|RNF 156|RNF156
- Storage Buffer:PBS, 0.1% sodium azide, 50% glycerol, pH 7.3.
- Storage Temperature:Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:RNF156
- Purification:Antigen affinity purified
- Packaging:Vial
Type:
Antigen: MGRN1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat