Anti-GCLC Rabbit Recombinant Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
Recommended dilution: IHC: 1:20 to 1:200
Type:
Antigen: GCLC
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77738-069
- 77855-858
- Antigen Symbol:
- GCLC
- GCLC
- Antigen Synonyms:
- EC 6.3.2.2|Gamma ECS|Gamma glutamylcysteine synthetase|Gamma-ECS|Gamma-glutamylcysteine synthetase|GCL|GCS|GCS heavy chain|GLCL|GLCLC|Glutamate cysteine ligase catalytic subunit|Glutamate--cysteine ligase catalytic subunit|GSH1_HUMAN
- EC 6.3.2.2|Gamma ECS|Gamma glutamylcysteine synthetase|Gamma-ECS|Gamma-glutamylcysteine synthetase|GCL|GCS|GCS heavy chain|GLCL|GLCLC|Glutamate cysteine ligase catalytic subunit|Glutamate--cysteine ligase catalytic subunit|GSH1_HUMAN
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- P48506
- P48506
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Immunogen:
- Recombinant Human Glutamate-cysteine ligase catalytic subunit protein (78-236AA)
- Recombinant Human Glutamate-cysteine ligase catalytic subunit protein (78-236AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice