Anti-KCNA1 Rabbit Recombinant Polyclonal Antibody (HRP (Horseradish Peroxidase))
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: IER2
Clonality: Recombinant Polyclonal
Clone:
Conjugation: FITC
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77858-398
- 77741-437
- Antigen Symbol:
- KCNA1
- KCNA1
- Antigen Synonyms:
- AEMK|EA1|Episodic ataxia with myokymia|HBK1|HUK1|Kca1 1|KCNA1_HUMAN|Kcpvd|KV1.1|MBK1|mceph|MGC124402|MGC126782|MGC138385|MK1|MK1, mouse, homolog of KV1.1|Potassium channel protein 1|Potassium voltage gated channel shaker related subfamily member 1|Potassium voltage gated channel subfamily A member 1|Potassium voltage gated channel, shaker related subfamily, member 1 (episodic ataxia with myokymia)|Potassium voltage-gated channel subfamily A member 1|RBK1|RCK1|Shak|Shaker related subfamily member 1|Voltage gated potassium channel subunit Kv1.1|Voltage-gated K(+) channel HuKI|Voltage-gated potassium channel HBK1|Voltage-gated potassium channel subunit Kv1.1
- AEMK|EA1|Episodic ataxia with myokymia|HBK1|HUK1|Kca1 1|KCNA1_HUMAN|Kcpvd|KV1.1|MBK1|mceph|MGC124402|MGC126782|MGC138385|MK1|MK1, mouse, homolog of KV1.1|Potassium channel protein 1|Potassium voltage gated channel shaker related subfamily member 1|Potassium voltage gated channel subfamily A member 1|Potassium voltage gated channel, shaker related subfamily, member 1 (episodic ataxia with myokymia)|Potassium voltage-gated channel subfamily A member 1|RBK1|RCK1|Shak|Shaker related subfamily member 1|Voltage gated potassium channel subunit Kv1.1|Voltage-gated K(+) channel HuKI|Voltage-gated potassium channel HBK1|Voltage-gated potassium channel subunit Kv1.1
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- HRP (Horseradish Peroxidase)
- HRP (Horseradish Peroxidase)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q09470
- Q09470
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Immunogen:
- Recombinant Human Potassium voltage-gated channel subfamily A member 1 protein (7-150AA)
- Recombinant Human Potassium voltage-gated channel subfamily A member 1 protein (7-150AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice