Anti-ERAP1 Rabbit Recombinant Polyclonal Antibody (HRP (Horseradish Peroxidase))
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: ERAP1
Clonality: Recombinant Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77736-074
- 77854-866
- Antigen Symbol:
- ERAP1
- ERAP1
- Antigen Synonyms:
- A LAP|A-LAP|Adipocyte derived leucine aminopeptidase|Adipocyte-derived leucine aminopeptidase|ALAP|Aminopeptidase PILS|Aminopeptidase regulator of TNFR1 shedding|APPILS|ARTS 1|ARTS-1|Arts1|Endoplasmic reticulum aminopeptidase 1|Endoplasmic reticulum aminopeptidase associated with antigen processing|ERAAP|ERAAP1|ERAP 1|ERAP1_HUMAN|KIAA0525|kiaa0525 protein|PILS AP|PILS-AP|PILSA|PILSAP|Puromycin insensitive leucyl specific aminopeptidase|Puromycin-insensitive leucyl-specific aminopeptidase|Type 1 tumor necrosis factor receptor shedding aminopeptidase regulator|type 1 tumor necrosis factor receptors shedding aminopeptidase regulator|VEGF-induced aminopeptidase
- A LAP|A-LAP|Adipocyte derived leucine aminopeptidase|Adipocyte-derived leucine aminopeptidase|ALAP|Aminopeptidase PILS|Aminopeptidase regulator of TNFR1 shedding|APPILS|ARTS 1|ARTS-1|Arts1|Endoplasmic reticulum aminopeptidase 1|Endoplasmic reticulum aminopeptidase associated with antigen processing|ERAAP|ERAAP1|ERAP 1|ERAP1_HUMAN|KIAA0525|kiaa0525 protein|PILS AP|PILS-AP|PILSA|PILSAP|Puromycin insensitive leucyl specific aminopeptidase|Puromycin-insensitive leucyl-specific aminopeptidase|Type 1 tumor necrosis factor receptor shedding aminopeptidase regulator|type 1 tumor necrosis factor receptors shedding aminopeptidase regulator|VEGF-induced aminopeptidase
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- HRP (Horseradish Peroxidase)
- HRP (Horseradish Peroxidase)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q9NZ08
- Q9NZ08
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Immunogen:
- Recombinant human Endoplasmic reticulum aminopeptidase 1 protein (642-941AA)
- Recombinant human Endoplasmic reticulum aminopeptidase 1 protein (642-941AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice