Anti-HOXA1 Rabbit Polyclonal Antibody
Catalog # 77737-964
Supplier: AFG Bioscience
New Product
Specifications
- Antigen Symbol:HOXA1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Mouse,Rat
- Western Blot:Yes
- Size:50 µg
- Form:Liquid
- Gene ID:P49639
- Antigen Synonyms:BSAS|Homeo box A1|Homeobox 1F|Homeobox A1|Homeobox protein Hox A1|Homeobox protein Hox-1F|Homeobox protein Hox-A1|Hox 1.6 like protein|Hox 1F|HOX A1|HOX A1 homeodomain protein|HOX1|HOX1F|hoxb1b|HXA1_HUMAN|Lab like protein|MGC45232
- Storage Buffer:PBS, 50% glycerol, 0.5% BSA, 0.02% sodium azide, pH 7.4
- Storage Temperature:Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:BSAS
- Purification:Antigen affinity purified
- Packaging:Vial
Specifications
About this item
Recommended dilution: WB:1:500 to 1:2000, IHC:1:100 to 1:300, ELISA:1:20000
Type:
Antigen: HOXA1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse;Rat