Anti-NR2C2AP Rabbit Recombinant Polyclonal Antibody (Biotinylated)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: NR2C2AP
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Biotinylated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77744-942
- 77744-941
- Antigen Symbol:
- NR2C2AP
- NR2C2AP
- Antigen Synonyms:
- nr2c2ap|NR2CA_HUMAN|Nuclear receptor 2C2 associated protein|Nuclear receptor 2C2-associated protein|Repressor for TR4 transactivation|TR4 orphan receptor associated 16 kDa protein|TR4 orphan receptor associated protein TRA16|TR4 orphan receptor-associated 16 kDa protein
- nr2c2ap|NR2CA_HUMAN|Nuclear receptor 2C2 associated protein|Nuclear receptor 2C2-associated protein|Repressor for TR4 transactivation|TR4 orphan receptor associated 16 kDa protein|TR4 orphan receptor associated protein TRA16|TR4 orphan receptor-associated 16 kDa protein
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Biotin
- Biotin
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q86WQ0
- Q86WQ0
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Immunogen:
- Nuclear receptor 2C2-associated protein (7-139AA)
- Nuclear receptor 2C2-associated protein (7-139AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage Temperature:
- Upon receipt, store at –20 °C or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 °C or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice