Anti-MDH2 Rabbit Recombinant Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
Recommended dilution: WB: 1:500 to 1:2000, IHC: 1:20 to 1:200
Type:
Antigen: MDH2
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77742-929
- 77859-607
- Antigen Symbol:
- MDH2
- MDH2
- Antigen Synonyms:
- M MDH|Malate dehydrogenase 2|NAD (mitochondrial)|Malate dehydrogenase|mitochondrial|MDH|MDHM_HUMAN|MGC:3559|Mitochondrial malate dehydrogenase 2|NAD|Mor 1|MOR1
- M MDH|Malate dehydrogenase 2|NAD (mitochondrial)|Malate dehydrogenase|mitochondrial|MDH|MDHM_HUMAN|MGC:3559|Mitochondrial malate dehydrogenase 2|NAD|Mor 1|MOR1
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- P40926
- P40926
- Isotype:
- IgG
- IgG
- Western Blot:
- Yes
- Yes
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Immunogen:
- Recombinant Human Malate dehydrogenase, mitochondrial protein (59-338AA)
- Recombinant Human Malate dehydrogenase, mitochondrial protein (59-338AA)
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage Buffer:
- PBS, 0.02% sodium azide, 50% glycerol, pH 7.3.
- PBS, 0.02% sodium azide, 50% glycerol, pH 7.3.
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice