Anti-APLF Rabbit Polyclonal Antibody
77736-502
New Product
- Antigen Symbol:APLF
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Mouse
- Western Blot:Yes
- Size:50 µg
- Form:Liquid
- Gene ID:Q8IW19
- Antigen Synonyms:RGD1565557|APLF_HUMAN|PNK and APTX like FHA protein|2010301N04Rik|AI452191|Aprataxin and PNK-like factor|C2orf13|PNK and APTX-like FHA domain-containing protein|XIP1|XRCC1 interacting protein 1|Apurinic-apyrimidinic endonuclease XRCC1-interacting protein 1
- Storage Buffer:PBS, 50% glycerol, 0.5% BSA, 0.02% sodium azide, pH 7.4
- Storage Temperature:Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:2010301N04Rik
- Purification:Antigen affinity purified
- Packaging:Vial
Recommended dilution: WB: 1:500 to 1:2000, IHC: 1:100 to 1:300, ELISA: 1:40000
Type:
Antigen: APLF
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse