Anti-GTPBP2 Rabbit Polyclonal Antibody
100 µl, Vial
Catalog # 77739-257
Supplier: AFG Bioscience
Supplier #: A24168-100UL
Jump to:
- Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
- Return Policy
Product Details & Documents
Type:
Antigen: GTPBP2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse;Rat
Specifications
- Size:100 µl
- Packaging:Vial
- Clonality:Polyclonal
- Purification:Antigen affinity purified
- Form:Liquid
- Storage Buffer:PBS, 0.1% sodium azide, 50% glycerol, pH 7.3
- Isotype:IgG
- Host:Rabbit
- Cat. No.:77739-257
- Reactivity:Human|Mouse|Rat
- Western Blot:Yes
- Shipping Temperature:Shipped on blue ice
- Antigen Symbol:GTPBP2
- Gene ID:Q9BX10
- Immunogen:RP11-22I24.2
- ImmunoChemistry:Yes
- Conjugation:Unconjugated
- Antigen Synonyms:GTP binding protein 2|GTP-binding protein 2|GTPB2_HUMAN|GTPBP 2|MGC74725|RP11-22I24.2
- ELISA:Yes
- Storage Temperature:Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.



