Anti-NDUFB8 Rabbit Recombinant Polyclonal Antibody (HRP (Horseradish Peroxidase))
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: NDUFB8
Clonality: Recombinant Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77744-200
- 77744-201
- Antigen Symbol:
- NDUFB8
- NDUFB8
- Antigen Synonyms:
- ASHI|CI-ASHI|Complex I ASHI subunit|Complex I-ASHI|mitochondrial|NADH dehydrogenase (ubiquinone) 1 beta subcomplex 8 19kDa|NADH dehydrogenase [ubiquinone] 1 beta subcomplex subunit 8|NADH dehydrogenase [ubiquinone] 1 beta subcomplex subunit 8 mitochondrial|NADH-ubiquinone oxidoreductase ASHI subunit|NDUB8_HUMAN|NDUFB8
- ASHI|CI-ASHI|Complex I ASHI subunit|Complex I-ASHI|mitochondrial|NADH dehydrogenase (ubiquinone) 1 beta subcomplex 8 19kDa|NADH dehydrogenase [ubiquinone] 1 beta subcomplex subunit 8|NADH dehydrogenase [ubiquinone] 1 beta subcomplex subunit 8 mitochondrial|NADH-ubiquinone oxidoreductase ASHI subunit|NDUB8_HUMAN|NDUFB8
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- HRP (Horseradish Peroxidase)
- HRP (Horseradish Peroxidase)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- O95169
- O95169
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Immunogen:
- NADH dehydrogenase [ubiquinone] 1 beta subcomplex subunit 8, mitochondrial protein (29-132AA)
- NADH dehydrogenase [ubiquinone] 1 beta subcomplex subunit 8, mitochondrial protein (29-132AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, protein G purified
- >95%, protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice