Anti-MRPS22 Rabbit Recombinant Polyclonal Antibody
New Product
Recommended dilution: IHC: 1:20 to 1:200
Type:
Antigen: MRPS22
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
- Catalog No:
- 77743-465
- 77859-994
- Antigen Symbol:
- MRPS22
- MRPS22
- Antigen Synonyms:
- 28S ribosomal protein S22|C3orf5|COXPD5|GIBT|GK002|mitochondrial|Mitochondrial ribosomal protein S22|MRP-S22|RPM S22|RPMS22|RT22_HUMAN|S22mt
- 28S ribosomal protein S22|C3orf5|COXPD5|GIBT|GK002|mitochondrial|Mitochondrial ribosomal protein S22|MRP-S22|RPM S22|RPMS22|RT22_HUMAN|S22mt
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- P82650
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Immunogen:
- Recombinant Human 28S ribosomal protein S22, mitochondrial protein (1-360AA)
- Recombinant Human 28S ribosomal protein S22, mitochondrial protein (1-360AA)
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage Buffer:
- PBS, 0.02% sodium azide, 50% glycerol, pH 7.3.
- PBS, 0.02% sodium azide, 50% glycerol, pH 7.3.
- Storage Temperature:
- Upon receipt, store at –20 °C or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 °C or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice