Anti-FARP2 Rabbit Recombinant Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: FARP2
Clonality: Recombinant Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77736-860
- 77855-201
- Antigen Symbol:
- FARP2
- FARP2
- Antigen Synonyms:
- FARP2_HUMAN|FERM|FERM domain including RhoGEF|RhoGEF and pleckstrin domain protein 2|RhoGEF and pleckstrin domain-containing protein 2|FGD1 related Cdc42 GEF|FIR|FRG|KIAA0793|PH domain-containing family C member 3|Pleckstrin homology domain-containing family C member 3|PLEKHC3
- FARP2_HUMAN|FERM|FERM domain including RhoGEF|RhoGEF and pleckstrin domain protein 2|RhoGEF and pleckstrin domain-containing protein 2|FGD1 related Cdc42 GEF|FIR|FRG|KIAA0793|PH domain-containing family C member 3|Pleckstrin homology domain-containing family C member 3|PLEKHC3
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- FITC (Fluorescein Isothiocyanate)
- FITC (Fluorescein Isothiocyanate)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- O94887
- O94887
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Immunogen:
- Recombinant human FERM, ARHGEF and pleckstrin domain-containing protein 2 protein (261-550AA)
- Recombinant human FERM, ARHGEF and pleckstrin domain-containing protein 2 protein (261-550AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice