Anti-MAGEA2 Rabbit Recombinant Polyclonal Antibody (HRP (Horseradish Peroxidase))
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: MAGEA2
Clonality: Recombinant Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77742-557
- 77859-304
- Antigen Symbol:
- MAGEA2
- MAGEA2
- Antigen Synonyms:
- MAGE2|MAGEA2A||MAGEA2B|Melanoma-associated antigen 2|Cancer/testis antigen 1.2|CT1.2|MAGE-2 antigen
- MAGE2|MAGEA2A||MAGEA2B|Melanoma-associated antigen 2|Cancer/testis antigen 1.2|CT1.2|MAGE-2 antigen
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- HRP (Horseradish Peroxidase)
- HRP (Horseradish Peroxidase)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- P43356
- P43356
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Immunogen:
- Recombinant Human Melanoma-associated antigen 2 protein (1-104AA)
- Recombinant Human Melanoma-associated antigen 2 protein (1-104AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage Temperature:
- Upon receipt, store at –20 °C or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 °C or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice