Anti-MT-CO1 Rabbit Recombinant polyclonal Antibody (HRP (Horseradish Peroxidase))
New Product
Type:
Antigen: MT-CO1
Clonality: Recombinant polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
- Catalog No:
- 77860-067
- 77743-559
- Antigen Symbol:
- MT-CO1
- MT-CO1
- Antigen Synonyms:
- COI|COX I|COX1|COX1_HUMAN|COXI|Cytochrome c oxidase polypeptide I|Cytochrome c oxidase subunit 1|Cytochrome C Oxidase subunit I|Mitochondrially encoded cytochrome c oxidase I|MT CO1|MTCO 1|MTCO1
- COI| COX I| COX1| COX1_HUMAN| COXI| Cytochrome c oxidase polypeptide I| Cytochrome c oxidase subunit 1| Cytochrome C Oxidase subunit I| Mitochondrially encoded cytochrome c oxidase I| MT CO1| MTCO 1| MTCO1
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Unconjugated
- HRP (Horseradish Peroxidase)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- P00395
- P00395
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Immunogen:
- Recombinant Human Cytochrome c oxidase subunit 1 protein (474-513AA)
- Recombinant Human Cytochrome c oxidase subunit 1 protein (474-513AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at −20 or −80°C, avoid repeated freeze
- Upon receipt, store at −20 or −80 °C, avoid repeated freeze
- Shipping Temperature:
- Shipped on Blue ice
- Shipped on Blue ice