Anti-MPZL2 Rabbit Recombinant Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
Recommended dilution: IHC: 1:20 to 1:200
Type:
Antigen: MPZL2
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77743-360
- 77859-925
- Antigen Symbol:
- MPZL2
- Antigen Synonyms:
- MPZL2|EVA|EVA1|UNQ606/PRO1192|Myelin protein zero-like protein 2|Epithelial V-like antigen 1
- MPZL2|EVA|EVA1|UNQ606/PRO1192|Myelin protein zero-like protein 2|Epithelial V-like antigen 1
- Clonality:
- Recombinant Polyclonal
- Conjugation:
- Unconjugated
- Reactivity:
- Human
- Host:
- Rabbit
- Gene ID:
- O60487
- O60487
- Isotype:
- IgG
- ELISA:
- Yes
- ImmunoChemistry:
- Yes
- Immunogen:
- Recombinant Human Myelin protein zero-like protein 2 protein (30-140AA)
- Recombinant Human Myelin protein zero-like protein 2 protein (30-140AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at –20 °C or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 °C or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice